VWR® Critical Swab® Swab, Micro Foam Head
Proveedor: VWR
VWR® Critical Swab®
TX821V
89023-000CS 8344.63 MXN
89023-000
VWR® Critical Swab® Swab, Micro Foam Head
Hisopos Hisopos para muestras
Swab features 100ppi open cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam head
  • Virgin polypropylene handle
  • No adhesive
Swab head is thermally bonded to the handle using no adhesive.