Puritan® Wooden Applicator/Stirring Sticks, Puritan Medical Products
Proveedor: Puritan Medical Products
Puritan®
807 25-8072W 25-8072W 805 809 805 809 807
10805-018CS 5263.6 MXN
87006-044 82030-614 CA82030-606 82030-606 CA82030-616 CA10805-018 CA82030-614 10805-018
Puritan® Wooden Applicator/Stirring Sticks, Puritan Medical Products
Imánes de agitación Varillas agitadoras
Wooden applicator sticks are made for general purpose functionality.
Choose a size and shape that works best for required applications, such as specimen transfer, mixing rods, and laboratory work. Sticks are manufactured from northern maine white birch, with straight cut ends.
Información para pedidos: Available in sterile or nonsterile formats. Sterile items are individually wrapped, with two sticks per package.