Lexa™ Splash GoggleGear™ Series Safety Goggles, 3M™ Company
Proveedor: 3M
Lexa™
7010045731
47746-326CS 45534.48 MXN
47746-326
Lexa™ Splash GoggleGear™ Series Safety Goggles, 3M™ Company
Gafas /Googles
GoggleGear™ Safety Goggles go beyond the standards with a combination of design and performance features rarely found with other safety eyewear.
  • Rubber flange provides comfort and a complete seal around perimeter
  • Fully assembled goggles with soft flange seals
  • Vented channels to increase airflow; minimize fogging and keep out particulates
  • Adjustable headband ensures a secure fit, even in active applications
  • High wrap design allows for a wide field of view
The goggle's soft flange seals the entire lens perimeter to keep out liquid splash, and includes indirect vent channels to increase airflow and minimize fogging. Product is fully assembled and comes with an adjustable headband for a secure fit in active applications.
They feature a lightweight frame, a high wrap lens, low-profile design, and a comfortable fit.
Certificaciones: Meets ANSI Z87.1-2010, CSA Z94.3-2007