Nunc® OmniTray Single-Well Plates with Lids, Thermo Scientific
Proveedor: Thermo Scientific
Nunc®
242811 264728 165218 264728 242811 242811 140156 140156 165218
46600-638CS 9850 MXN
73520-160 CA73520-160 CA62409-600 62409-600 CATX62409-600 46600-638 CA46600-638 CA62409-590 62409-590
Nunc® OmniTray Single-Well Plates with Lids, Thermo Scientific
Microplacas Microplacas HTS
Designed for many applications, including high throughput screening
These polystyrene, single-well plates have the same footprint as a 96-well plate. They also hold a 11.4x7.3cm (41/2x27/8") nylon membrane used for dot blotting bacterial clones. Lid protects cells from contamination while providing a means for cell respiration during incubation and/or storage. Radiation sterilized.
Nunclon™Δ certified surface ensures cell attachment and growth. MaxiSorp™ surface is ideal for arraying molecules with mixed hydrophilic/hydrophobic domains, such as proteins, directly onto the plate. Plate featuring an enhanced rectangular area is ideal for automatic picking and viewing systems.
Plates are useful for screening DNA libraries in high volume laboratories using robotics. They are also suitable for manual plating and screening. Compatible with Beckman Coulter®, Biomek®, Hybaid, Orca, Tecan® Trak, and Zymark® robotic systems.
Información para pedidos: Plates packaged 10 sleeves per case.