PIG® SpillBlocker® Tabletop Dike, New Pig
Proveedor: New Pig
PIG® SpillBlocker®
PLR229
46000-742EA 4300 MXN
46000-742
PIG® SpillBlocker® Tabletop Dike, New Pig
Materiales para contención de derrames
Contains benchtop spills without absorbing; Excellent for small spills on smooth work surfaces.
  • Polyurethane material resists water, oils and many chemicals
  • High-visibility yellow color draws attention to spill for increased safety
  • Reusable dike cleans easily with soap and water
  • Comes in compact storage box for easy transport
  • Cut to any length for a custom fit
Can also be used to form an edge on workbenches to keep bolts, bearings and other small parts from rolling off.