Nalgene® Desiccator Cabinets, Acrylic, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
5317-0120 5317-0180 5317-0070
24987-056EA 36806.4 MXN
24987-056 24987-050 24987-053
Nalgene® Desiccator Cabinets, Acrylic, Thermo Scientific
Desecadores Armarios desecadores
For storing anhydrous and hygroscopic biological and chemical compounds, as well as electronic assemblies and any components requiring controlled storage conditions.
Provide dust- and moisture-free storage in refrigerators and freezers, and on the benchtop. Neoprene gasket on door ensures a tight seal. Removable, lipped tray holds solid desiccant. Models are available with 2, 3, and 4 shelves with adjustable grooves for 3, 5, and 7 positions, respectively. Shelves are removable, adjustable, and vented to maximize air flow.