SP Bel-Art Secador® 1.0, 2.0, 3.0, 4.0 Vertical Desiccator Cabinets, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a part of SP
Secador®
F42074-1000 F42071-0006 F42073-1008 F42074-1008 F42073-1000 F42071-0007 F42072-1000 F42071-0000 F42074-1007 F42074-1006 F42071-0008 F42072-1008
47750-986EA 23892.01 MXN
47750-986 47743-424 47743-446 89080-014 47743-442 47743-420 89080-010 47743-422 47743-444 89080-008 89080-018 47750-984
SP Bel-Art Secador® 1.0, 2.0, 3.0, 4.0 Vertical Desiccator Cabinets, Bel-Art Products, a part of SP
Desecadores Armarios desecadores
The attractive design and sturdy construction of the Secador® line of desiccator cabinets assures easy and reliable storage of moisture- and light-sensitive products.
  • Sturdy construction
  • Easy and reliable storage
  • Protects moisture- and UV-sensitive products
  • Co-polyester construction
Choose from an extensive selection of sizes, styles and colors of desiccator cabinets to meet your specific dry storage needs
Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals. Perforated shelves facilitate circulation of dry air and a space for the desiccant is under the bottom shelf. An easy-to-view digital hygrometer is located in the door front.
1.0, 2.0, 3.0 and 4.0 vertical units have a compact 34.1W x 41.4 cmD footprint for space saving stacking up to 3 units high and a side swinging door that opens to the left. 4.0 horizontal units have a wider footprint, lower height and pull down “oven style” door. Replacement shelves are available and the 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.
Durastar® co-polyester construction blocks 99% of UV light* and is resistant to staining, crazing and chemical attack. Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds.
Información para pedidos: All units can be used with certain desiccant cartridges, contact your Avantor representative for more information. U.S. Patents: 6,834,920; 7,318,630 and D474,626.