Mantener la sesión abierta?
Finalizando en 
Su sesión ha finalizado
Su sesión ha caducado. Por seguridad lo hemos desconectado.
¿Quiere conectarse de nuevo?
Uvex Genesis® Protective Eyewear, Honeywell Safety
Proveedor: HONEYWELL SAFETY PRODUCTS USA INC.
uvex genesis® Genesis®
S3221X S3202 S3240X S3203 S3240X S3211X S3221 S3240 S3200X S3223 S3210X S3204 S3240 S3201X S3244 S3201X S3221 S3241X S3224 S3211X S3203 S3220X S3200D S3200X S3202 S3220 S3200D S3220X S3260X S3220X S3260X S3200 S3244 S3221X S3213X S3260 S3211X S3223 S3200 S3260X S3241X S3213X S3204 S3243 S3260 S3224 S3220 S3210X S3243
94023-916EA 354.32 MXN InStock
15001-588 CA15001-576 15001-612 15001-578 CA15001-612 14216-000 15001-586 15001-610 15001-570 15001-592 14216-002 15001-580 CA15001-610 CA15001-574 CA15001-622 15001-574 CA15001-586 CA15001-616 CA15001-594 14216-001 CA15001-578 CA15001-584 94023-916 CA15001-570 15001-576 15001-582 CA94023-916 15001-584 CA14227-817 CA-15001-584 14227-817 CA15001-568 15001-622 CA15001-588 CA14231-228 14227-815 CA14216-000 CA15001-592 15001-568 CA14227-816 15001-616 14231-228 CA15001-580 15001-620 CA14227-815 15001-594 CA15001-582 CA14216-002 CA15001-620
Uvex Genesis® Protective Eyewear, Honeywell Safety
Gafas
Designed for comfort and compliance, Uvex Genesis® eyewear combines a lightweight, resilient multi-material frame with a soft elastomer brow guard for unsurpassed impact resistance.
  • Dual 9-base wrap-around lens provides uncompromised peripheral vision and protection Elastomer brow-guard diffuses and deflects impact
  • Temple length and lens inclination are adjustable
  • Soft fingers conform to any nasal profile
  • Frames guaranteed for life
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
The dual 9-base wraparound polycarbonate lens provides uninterrupted peripheral coverage and visibility
Clear lenses are appropriate for most indoor applications. Gold mirror, amber, and espresso lenses minimize the effects of sunlight and glare. The SCT-Reflect 50 lenses are designed for use in work applications with varying light conditions. This tint is a comfortable solution for workers who move between shaded or indoor locations and direct sunlight. SCT-Vermillion lenses are ideal for indoor applications requiring contrast, such as inspection. SCT-Blue lenses help counteract sodium vapor lighting. The Patriot® RWB frame features a red, white, and blue design with miniature U.S. flags at temples and “United We Stand” inscribed on bottom right lens outside field of vision.
Eyewear meets military Vo ballistic test for impact and ANSI Z87+ and CSA Z94.3 standards.
Soft, flexible “fingers” adjust and conform to any nasal profile, minimizing slippage. A three-position ratchet lens inclination system completes the custom fit. A ventilation channel between frame and lens vents the eye socket area for outstanding moisture/fog control. Lens replacement is quick, easy, and economical. Ultra-dura® lens coating provides superior scratch and chemical resistance. Uvextreme® lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection. Dura-streme® lens coating features Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity.
Order Now
Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.
PEDIDO
ACCESORIOS
INFORMACIÓN RELACIONADA
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
O
Revise y compare las alternativas
Disponemos de alternativas con las que puede ahorrar hasta  por producto unitario.  Para comparar detalles de los productos, seleccione hasta 3 alternativas y haga clic en comparar seleccionados. Para añadir productos a su cesta, introduzca la cantidad y haga clic en Añadir a la cesta.
Producto Original:
Descripción Numero del catalogo Disponibilidad Unidad Your Price
(IVA no incluido)
Precio Por Uds.