VWR® Reusable/Disposable Desiccant Cartridges
Proveedor: VWR
VWR®
42048-6065
47744-624PK 1499.96 MXN
47744-624
VWR® Reusable/Disposable Desiccant Cartridges
Desecadores Desecantes
Reusable and disposable desiccant cartridges are useful anywhere desiccation is needed.
  • Contain inert, non-corrosive, and non-toxic silica gel
  • Highly porous membrane covers the dish providing excellent moisture absorption
  • Desiccant can be easily regenerated by heating cartridges in a 50°C (122°F) oven overnight
  • Fit “Space Saver” desiccators, in addition to other plastic desiccators of similar size
  • Individually foil sealed
Clear polystyrene dish allows instant visual inspection of silica gel, which changes in color from blue to pale pink as it absorbs moisture. For more rapid drying, cut the membrane, transfer the spent gel crystals into a glass or porcelain dish, and place in a 150°C (300°F) oven for one hour.