VWR® Hand Cream
Proveedor: Avantor
VWR®
999516
89005-468
VWR® Hand Cream
Limpieza de la piel Cremas Cremas de manos
Designed specifically for laboratory technicians who wash frequently.
  • Nongreasy skin moisturizer absorbs quickly upon application
  • Prevents irritation from latex gloves, harsh soaps, and other irritants
  • Fragrance free, petroleum free
Non-greasy skin moisturizer absorbs quickly upon application and provides a barrier that prevents irritation from latex gloves, harsh soaps, and other irritants. Fragrance-free and contains no petroleum products that would compromise latex integrity.