Crews® Klondike® Protective Eyewear, MCR Safety
Proveedor: MCR Safety
Crews®
KD113 KD119 KD119AF KD110AF KD11B KD112 KD110 KD112AF KD117 KD114
33500-836EA 91.48 MXN
33500-850 80081-949 80081-959 33500-842 33500-840 33500-846 33500-836 33500-844 33500-848 33500-838
Crews® Klondike® Protective Eyewear, MCR Safety
Gafas
Strong, lightweight, scratch-resistant polycarbonate lenses have an 11° base curve and filter out 99.9% of UV radiation.
  • Stylish black matte-finished frames
  • Gel nosepiece for long-wearing comfort
  • Full orbital seal with dual lens design
  • Soft, secure TPR nose pad
Adjustable temples ensure comfortable fit for both male and female wearers.
Certificaciones: Eyewear meets or exceeds ANSI Z87.1 standards.