TRIWEAR® Metal Pro Grade Protective Eyewear, MCR Safety
Proveedor: MCR Safety
TRIWEAR®
T3112AF T3119AF T3110AF
33500-922CS 50458.46 MXN
33500-918 33500-922 33500-916
TRIWEAR® Metal Pro Grade Protective Eyewear, MCR Safety
Gafas
Ideal protective glasses for extended wear.
  • Platinum frame and resilient spring-hinged temples
  • Soft, secure proprietary gel nosepiece promotes a comfortable extended wear
  • Strong, lightweight, scratch-resistant polycarbonate lenses
  • 99% UV protection
Aspheric lens design eliminates headache and distortion problems caused by poor optical fit.
Widest possible optically correct viewing zone, permitting 180° of clear vision regardless of the wearer’s interpupillary distance
Certificaciones: ANSI Z87.1
Información para pedidos: Each pair comes with a black satin carrying case that can be used as a cleaning cloth.