BX™ Magnifying Safety Glasses, 3M™
Proveedor: 3M
BX™
7000127494 7000127494 7000127490 7000127492 7000127493 7000127491 7000127493 7000052794 7000052794 7000127491 7000127492
89006-661CS 11790.24 MXN
CA89006-663 89006-633 89006-661 89006-663 CA89006-661 CA89006-659 CA89006-637 89006-635 CA89006-635 89006-637 89006-659
BX™ Magnifying Safety Glasses, 3M™
Gafas
BX™ protective reader eyewear is designed for workers who have difficulty reading small print or are engaged in small detail work.
  • Features magnifying diopters, lightweight lenses with adjustable soft tip temples, and integrated sideshields
  • Adjustable temple length, lens angle, and nose bridge to allow a more comfortable fit
  • Impact-resistant polycarbonate lenses absorb 99.9% UV rays
Eye protection is important to safeguard your eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. Comfortable and attractive eyewear increases the usage of safety glasses on the job.
The ability to customize the safety glasses to the wearer makes them more pleasant to use over extended time periods. The nose bridge is cushioned for softness and comfort. The sleek, contoured design offers an attractive appearance along with functional eye protection. These safety glasses are recommended for use in assembly and mechanical, chipping, chiseling, cleaning, drilling, grinding, machining, masonry, metal repair, painting, pouring/casting, power fastening, riveting, sanding, and sawing applications. Industries in which these safety glasses are commonly used include aviation, construction, food and beverage, general manufacturing, mining, oil and gas, primary metals, and transportation.
Certificaciones: Meets Standards CSA Z94.3-2007 and ANSI Z87.1-2010.