Uvex Millennia™ Protective Eyewear, Honeywell Safety
Proveedor: Honeywell Safety
uvex millennia™
11150350 11150354 11150370 11150353 11150350
32999-800CS 3232.15 MXN
32999-818 32999-809 32999-806 32999-800 CA32999-800
Uvex Millennia™ Protective Eyewear, Honeywell Safety
Gafas
Uvex Millennia™ safety eyewear combines futuristic wrap-around styling with exceptional coverage and protection. Comfortable and lightweight, the face-contouring frame provides exceptional brow and side protection for the most demanding work environments.
  • Wrap-around unilens design, comfortable universal nosebridge
  • High impact dual 9-base wrap-around unilens
  • Clear lenses
Available with an extensive selection of high-performance lens tints and coatings, the Uvex Millennia™ is a popular choice among workers.
One-piece lens features an anti-scratch coating. Glasses include a comfortable universal nosepiece and an adjustable break-away Flexi-Cord™ neck cord for added security and convenience.
Certificaciones: Meets ANSI Z87+ (High Impact) and tested by Honeywell to meet the requirements of the CSA Z94.3 standard. Meets military V0 ballistic test for impact.