VWR® Antimicrobial Laboratory Hand Soap-TS
24410-97
89005-478CS 5874.45 MXN
89005-478
VWR® Antimicrobial Laboratory Hand Soap-TS
This pleasant smelling hand soap is ideal for laboratory and healthcare personnel that are required to wash frequently.
  • Ideal for laboratory and healthcare personnel required to wash frequently
  • Fast-acting antimicrobial agent provides efficacy against wide variety of bacteria
  • pH-balanced formulation is gentle on the hands
Its fast acting antimicrobial agent (0.3% Triclosan) provides efficacy against a wide variety of gram-positive and gram-negative bacteria including yeast-fungi. The pH balanced formulation is gentle on the hands.