VWR® Hand Sanitizer Gel
Proveedor: VWR
VWR®
24410-95
89005-474CS 3682.55 MXN
89005-474
VWR® Hand Sanitizer Gel
Limpieza de la piel Desinfectantes de la piel Desinfectantes para la piel en gel
Effective against a broad spectrum of microorganisms.
  • Quick-Drying and Leaves No Sticky Residue
  • Ideal for Reducing Bacteria on Hands Prior to Gloving
  • Helps Moisturize Hands and Prevent Chapping
Non-drying, antimicrobial hand gel can be used anywhere soap and water are not available
Meets OSHA requirements for hand washing.
The quick-drying gel leaves no sticky residue and is ideal for reducing bacteria on hands prior to gloving. Special emollients in the 62% ethyl alcohol formula help moisturize hands and prevent chapping.