VWR® PCMX Antimicrobial Laboratory Hand Soap
Proveedor: Avantor
VWR®
24410-98 24410-96
89005-476CS 13701.02 MXN
89005-476 CA89005-480
VWR® PCMX Antimicrobial Laboratory Hand Soap
Limpieza de la piel Jabones y productos para la piel
Designed for laboratory workers who wash their hands frequently
  • Designed for laboratory workers who wash their hands frequently
  • Will not dry out the skin and replenishes natural oils
  • 1% chloroxylenol (PCMX) antimicrobial agent is effective against bacteria and fungi
The 1% chloroxylenol (PCMX) antimicrobial agent is effective against gram-positive and gram-negative bacteria as well as fungi.
Unique formulation will not dry out the skin, and replenishes natural oils that are washed away by other hand detergents.