Fuse™ Protective Eyewear, Honeywell Safety
Proveedor: Honeywell Safety
11150801 11150355 11150800 11150800 11150805
89030-543EA 347.1 MXN
89030-543 16003-872 16003-866 16003-868 CA16003-866
Fuse™ Protective Eyewear, Honeywell Safety
Gafas
A 9-base wraparound design provides a distortion-free 180° field of vision. The frame features a nickel-free topcoat. Lenses are made from optical-grade polycarbonate with a protective hardcoat to maximize the life of the eyewear. Glasses are available with Fog-Ban® antifog coating, and include an adjustable breakaway FlexiCord™ neck cord for added security and convenience.
  • Soft, adjustable saddle nose pads
  • Flexible temple tips for comfortable fit
  • Lightweight and durable
Certificaciones: Meets ANSI Z87+ and CSA Z94.3 standards.