Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Proveedor: Thermo Scientific
Nunc® MicroWell™
237105 237105 236105 236108 237107 237107 236105 236108
43300-434CS 11968.11 MXN
CA43300-438 CA43300-448 43300-438 43300-450 CA43300-450 43300-448 CA43300-434 43300-434
Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Microplacas Microplacas para inmunología
Black and white 96-well plates are specifically designed for fluorescence and luminescence assays
Nunclon™Δ treated surface ensures optimal cell adhesion and growth. Non-treated surface is ideal for use in liquid phase immunoassays, serological tests and other homogeneous assays. MaxiSorp™ surface has a high affinity for proteins, especially antibodies. PolySorp™ surface is less polar than MaxiSorp and has an affinity towards molecules with mainly hydrophobic domains. MaxiSorp™ and PolySorp™ surfaces are used in many solid phase immunoassays.
Made of polystyrene, they feature a flat-bottom well design and have a total well volume of 400µL with a recommended working volume of 200 to 350µL. Select resins and manufacturing methods ensure maximum signal, low background, and minimal signal crosstalk. Alphanumeric coordinates offer easy orientation and sample identification. Meet the proposed SBS industry guidelines.
Información para pedidos: Plates are packaged 10 per sleeve with the exception of 43300-434, which is packed 1 per sleeve.