GASTIGHT® 1000 Series Syringes, Hamilton
Proveedor: Hamilton
GASTIGHT®
81416 81616 81316 82527 81401 81514 81327 81516 7649-01 81517 81317 81301 81417 81617 81330 82520 81501 85027 85020 81643 81314 85021 86020 7650-01 81627 81443 81343 203270 81527 7651-01 81414 81365 203260 81530 81543 81630 81601 81430 81322 81427
60376-558EA 3007.53 MXN
60373-866 60373-868 89210-072 60376-438 60376-678 60376-558 60376-434 60376-412 60376-300 60376-565 10192-086 60376-321 60376-285 60376-561 60376-140 60376-680 10192-084 89210-074 60376-161 60373-870 72310-008 60375-624 60375-668 10192-116 60376-128 60376-106 10192-112 60376-423 10192-114 60376-674 82020-762 60376-650 60376-354 72310-392 82020-764 60373-862 60375-670 60373-864 60373-860 89221-472
GASTIGHT® 1000 Series Syringes, Hamilton
Jeringas Jeringas para cromatografía
The 1000 series is a mid volume Gastight® syringe. Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
This series of syringes is ideal for dispensing volumes from 100μL up to 100mL
A variety of different terminations and needle options are available. Syringe Terminations: Removable needle syringes are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. This termination also allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Luer tip syringes feature needles that are removable and fit over a ground glass hub which is tapered in the shape of a male luer. This termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination also increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
Luer tip cemented needle syringes are mid-volume syringes in which the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
Needle Point Styles: Point style 2 is a sharp, beveled, curved, non-coring needle point recommended for septum penetration. Point style 3 is a blunt needle point for use with HPLC injection valves and for sample pipetting. Point style 5 is a conical needle with side port for penetration of septa, thin-gauged vinyls, and plastics without coring.
Información para pedidos: For additional available needle points or gauges, contact your Avantor representative.