Visorgogs® Safety Goggles, Jones FR
Proveedor: Jones FR
Visorgogs®
34862-BLUE 34862-WHITE 34862-RED 34854 34862-GREEN
33012-008EA 319.15 MXN
33012-008 33012-005 33012-006 33012-007 33012-000
Visorgogs® Safety Goggles, Jones FR
Gafas /Googles
These full vision safety goggles are specially designed to protect eyes from flying particles or splashing liquids.
  • Indirect ventilation screen keeps dirt and dust out and keeps lenses fog-free
  • Lenses are designed strutless with a down-angle vision for clear visibility
  • Goggles can be worn over eyeglasses
  • Available in four visor colors with clear lenses
  • Replacement lens fits both indirect and direct vent goggles
Unique design incorporates comfortable, impact-resistant goggles with a vented, opaque visor to shade eyes and reduce eye strain and fatigue.
Certificaciones: Meet ANSI Z87.1 standard.