For low-viscosity liquids like aqueous solutions, plasma, and lightweight oils
Model 101 stirs 18.9L (5gal.) of aqueous solution at speeds from 500–7500rpm. Solid-state electronic voltage control varies speed steplessly over the full range. Internal fuse protects the motor from overload.
Model 102 has all the features of the 101 but adds a 10:1 reduction gear head for a second, lower speed range of 50–750rpm. Stirs up to 7.6L (2gal.) of a medium-weight oil at lower speeds.
Model 105, a constant speed stirrer set at 1550rpm, mixes 37.9–56.8L (10–15gal.) of aqueous solution. Nonsparking motor has on/off switch (no speed control). Internal thermal overload protection feature shuts off a hot motor. Can be retrofitted with a variable-speed controller, Model 115 Adjusta-Speed (58950-210).
Caution: Stirrers are not explosion-proof. Do not use with flammables.
Compact, rugged stirrers are built to withstand continuous use under normal loads. Each stirrer is supplied with a true-running, precision collet chuck designed to accept an accessory shaft with the same diameter as the motor shaft.
Información para pedidos: Complete with speed controller (except Model 105), grounded three-wire plug and cord, appropriate collet-type chuck, instruction manual, and supporting rod with wrench and carriage bolt for mounting. For mixing paddles, see 58958-405 series. Clamps and support stands sold separately; contact your VWR representative for more information.