J.T.Baker® NEUTRASORB®, Material for Acid Spills
Proveedor: Avantor
NEUTRASORB®
BAKR4456-05 BAKR4456-05
JT4456-5CS 29993.78 MXN
JT4456-5 CAJT4456-5
J.T.Baker® NEUTRASORB®, Material for Acid Spills
Materiales para contención de derrames
NEUTRASORB® material provides rapid, controlled neutralization of concentrated acids and visible color change as indication of complete neutralization
Caution: NEUTRASORB® material should not be used for spills of hydrofluoric acid, fuming nitric acid, fuming sulfuric acid, or peroxy (per-) organic or other highly reactive acids.
After complete neutralization, waste materials contain neutralized alkali metal and alkaline earth salts of the treated acid.
Información para pedidos: Available in three sizes or as part of a complete acid spill clean-up kit (see JT4433-2 series for kits).