Stools have a flexible, nonmarking 35.6 cm (14") diameter urethane seat, a 66 cm (26") diameter composite base with 5.1 cm (2") casters, and pneumatic seat adjustment.
- Flexible, nonmarking seat
- Urethane fabrication
- Pneumatic seat adjustment
Stools are easy to clean and resistant to stains and damage. Medium and high bench heights are equipped with a 50.8 cm (20") diameter adjustable height footring.
Información para pedidos: Contour seating is covered by a five-year warranty against defects in material, workmanship or abnormal wear. Upholstery is covered for one year. Pneumatic cylinders are covered for the life of the product.