Rubber Gas Expansion Bag, Walter Stern
Proveedor: Walter Stern
206
56335-003EA 737.91 MXN
56335-003
Rubber Gas Expansion Bag, Walter Stern
Muestreadores Muestreadores de gas
Two green latex rubber bulbs, each with approximately 200mL capacity
First bulb equipped with valves to provide a constant air stream to the second netted bulb, with 51cm (20") tubing. Inside diameter of tubing is 0.6cm (1/4").