SP Bel-Art® Long-Handled Dippers, Polyethylene, Bel-Art Products, a part of SP
Proveedor: Bel-Art
Scienceware®
36780-6016 36781-6032 36781-6016 36782-6016
13197-238EA 2997.24 MXN
13197-238 13197-236 13197-232 56920-060
SP Bel-Art® Long-Handled Dippers, Polyethylene, Bel-Art Products, a part of SP
Muestreadores Tomamuestras
High-density polyethylene dipper for easy sampling from tanks, vats, or lakes.
  • HDPE construction
  • Chemically resistant
  • Corrosion-resistant
  • Dipper is constructed of two screw-together pieces
Will not corrode or contaminate solutions
Dippers are available with 500 or 1000 ml capacities. The 3.7m (12') dipper is constructed of two screw-together pieces.
The 2.9 cm (11/8") diameter handle is welded to the rigid bowl at a 45° angle. With hook at the end of handle for convenient hanging.