VWR® Hydrometers for Liquids Considerably Heavier than Water
Proveedor: VWR
VWR®
61801-8511 61801-8411
34620-108EA 1085.78 MXN
34620-108 34620-110
VWR® Hydrometers for Liquids Considerably Heavier than Water
Hidrómetros
These hydrometers have varied specific gravity ranges for determining the specific gravity of liquids heavier than water.
  • For determining the specific gravity of liquids much heavier than water
  • Calibrated at 60°/60°F
Approximately 305mm (12") long.