Disposable Safety Scalpels, Sterile, Integra™ Miltex®
Proveedor: Integra LifeSciences
4-515C 4-515 4-510 4-511
21909-670PK 1013.52 MXN
95039-117 21909-674 21909-672 21909-670
Disposable Safety Scalpels, Sterile, Integra™ Miltex®
Escalpelos Escalpelos
Pre-assembled scalpels feature plastic handles and fully retractable stainless steel blades.
  • Size is clearly marked on each blade
  • Metric ruler on each scalpel assists in determining incision length
  • Five-year shelf life
Blades have a razor-sharp cutting edge for superior cutting and clean incisions.
Certificaciones: Scalpels comply with OSHA directive for the Needlestick Safety and Prevention Act.
Envase: Individually packaged in sealed, peel-apart pouches.