Bard-Parker® Rib-Back™ Carbon Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Proveedor: Aspen Surgical
Bard-Parker® Rib-Back™
371112 371115 371111 371110
89176-346CS 4810.51 MXN
89176-344 89176-346 89176-350 89176-348
Bard-Parker® Rib-Back™ Carbon Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Escalpelos Hojas de escalpelo
For use with No
3, No. 5, or No. 7 Bard-Parker® scalpel handles. Fine blade carbon steel. The Rib-Back™ is designed for greater cutting control and strength. Specially ground and carefully inspected. Blades individually packed in sterile foil pouch.