Sterile Scalpel Blades, Walter Stern
Proveedor: Walter Stern
320-003 320-001 320-005 320-002
25608-065PK 2041.44 MXN
25608-065 25608-087 14006-940 14006-938
Sterile Scalpel Blades, Walter Stern
Escalpelos Hojas de escalpelo
Fit handles 25607-925 and 25607-947
Made of high-grade vacuum hardened steel with excellent cutting ability. Sterilized by gamma radiation. Packed individually in aluminum foil.