Disposable Scalpel Blades, Sterile, Integra™ Miltex®
Proveedor: Integra LifeSciences
4-112B 4-121 4-312B 4-315 4-323 4-115 4-110 4-115C 4-315C 4-120 4-125 4-312 4-111 4-322 4-112 4-123 4-122 4-311 4-325 4-321 4-320 4-310
21909-628PK 2636.18 MXN
21909-618 21909-628 21909-616 21909-638 21909-648 21909-626 21909-610 21909-620 21909-642 21909-378 21909-630 21909-652 21909-614 21909-624 21909-646 21909-612 21909-622 21909-644 21909-382 21909-380 21909-650 21909-384
Disposable Scalpel Blades, Sterile, Integra™ Miltex®
Escalpelos Hojas de escalpelo
Carbon steel and stainless steel surgical blades feature razor-sharp cutting edges for superior cutting and clean incisions
Blades are for use with No. 3 or No. 4 scalpel handles. No. 3 scalpel handles accommodate blade numbers 10 through 15C. No. 4 scalpel handles accommodate blade numbers 20 through 25. Size is clearly marked on each blade. Individually packaged in sealed, peel-apart pouches. Five-year shelf life.