Bard-Parker® Scalpel Handle, No. 4, Aspen Surgical
Proveedor: Aspen Surgical
Bard-Parker®
371040
25859-000CS 4018.03 MXN
25859-000
Bard-Parker® Scalpel Handle, No. 4, Aspen Surgical
Escalpelos Mangos de escalpelos
For use with No
20, 21, 22, or 23 Bard-Parker® Rib-Back™ blades.