Designed to produce electronically adjustable intensity levels to meet the most exacting requirements
Heat shield accessory protects objects from infra-red energy (heat) emitted from bulbs. IR rejection rate is about 90%. Mounting clips included.
Feature neon power on/off indicator, long life replaceable reflector bulbs and a weighted base. Also feature durable chrome, high-gloss enamel on steel, and anodized aluminum bulb guards. All-angle swivel and 11" long chrome flex arm create adjustable light heads. A built-in socket for the magnifier accessory kit allows the lamps to be used to see “mini” to “macro” sizes. Gray base is 11/2" high with a 53/4" diameter. Illuminators operate on 120V, 50/60Hz.
Información para pedidos: One-year parts and labor warranty. Supplied with 6-foot line cord with 3-wire plug. Single head illuminators are supplied with one bulb; dual head illuminators are supplied with two bulbs. Dyna-Lite illuminators are supplied with tungsten/quartz/halogen combination beam bulb(s). Sun-Lite I illuminators are supplied with tungsten/argon spot beam bulb(s). All units can be ordered to 220 VAC upon request. Contact your VWR Sales Representative for information.