X-ACTO® No. 1 Precision and Safety Grip Knives, Elmer's
Proveedor: Elmer's
X-ACTO®
X3001 X3201
55412-026CS 1376.9 MXN
55412-044 55412-026
X-ACTO® No. 1 Precision and Safety Grip Knives, Elmer's
Cuchillos
For delicate, precision cutting, trimming, and stripping. Knives cut paper, plastic, balsa, thin metal, cloth, film, and acetate.
  • Precision knives feature a lightweight handle (No. 11 classic)
  • Available in a boxed or carded version
Información para pedidos: Knives with safety cap are boxed; knife without cap is carded.