UVP Mini UV Lamp, Analytik Jena
Proveedor: Analytik Jena
95-0305-03 95-0188-02 95-0158-04 95-0125-05
82027-150EA 1237.32 MXN
82027-154 36582-048 82027-150 82027-148
UVP Mini UV Lamp, Analytik Jena
Luces Lámparas de rayos UV
These lamps are excellent for quick field inspection or fluorescence applications.
  • For Field Use and Fluorescence Work
  • Longwave, Shortwave, and Combination Options Available
  • Four-Watt Intensity
  • Impact-Resistant Plastic
Mini UV Lamps feature four-watt intensity and are powered by four AA batteries (not included)
Each lamp housing is made of impact-resistant plastic that resists fading or cracking. The lamps are lightweight and small enough to fit into a pocket. A nylon strap is attached for easy handling.
The lamps are available in three configurations: longwave, shortwave, or a multi-band longwave/shortwave combination.
Información para pedidos: Model UVL-4F includes a flashlight and a longwave UV.