Mortar and Pestle Sets, Glass, Variety Glass
Proveedor: Variety Glass
826-2 826-6F 826-5 826-3 826-4
50415-100CS 17209.68 MXN
50415-100 50415-085 50415-041 50415-063 50415-020
Mortar and Pestle Sets, Glass, Variety Glass
Morteros y manos de mortero Morteros con manos de mortero
Mortar and pestle sets are made of clear, heavy, flint glass
Mortar has a heavy molded base and lip for pouring. Sold as a set.