SP Bel-Art® Ellipso-Spoon® and Spatula Samplers, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a part of SP
Ellipso-Spoon®
H36809-0040 H36807-0025 H36807-0021 H36808-0030 H36809-0050 H36806-0018 H36806-0015
70690-178CS 10105.05 MXN
70690-178 13197-398 13197-394 47727-724 13197-392 70680-298 13197-390
SP Bel-Art® Ellipso-Spoon® and Spatula Samplers, Bel-Art Products, a part of SP
Muestreadores Muestreadores de sólidos
This utensil’s specially curved shape permits contact with all corners of bottles and containers, virtually eliminating material or sample loss.
  • Curved shape improves sample collection
  • Easily retrieve solid and liquid samples
  • Polished stainless steel construction
Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container.
The Ellipso-Spoon® Sampler has a spoon end and a spatula end, and is made of polished 304 grade stainless steel.