VWR® Flexible-Arm™ Lights
Proveedor: VWR
VWR® Flexible Arm™
3095 3096 3098
23609-166EA 3136.08 MXN
23609-162 89030-200 23609-166
VWR® Flexible-Arm™ Lights
Luces Lámparas incandescentes
These lights are ideal for inspection, quality control, electronics, and sample examination.
  • Place light exactly where you need it
  • Easily maneuverable
Fingertip control raises, lowers, and pivots the perfectly balanced lamp wherever desired. Weighted metal base and spring counterbalance permit smooth, fluid movement with excellent stability. Beam closely resembles daylight for correct color detection.
Certificaciones: UL listed.
Información para pedidos: Supplied with white reflector and a 35W halogen bulb.