ERGOMAT® Nitril ESD-Conductive Ergonomic Floor Mats, ERGOMAT
Proveedor: Ergomat
Ergomat®
ENC0210 ENC0414 ENC0214 ENC0204 ENC0314 ENC0410 ENC0219 ENC0208 ENC0319 ENC0215 ENC0413 ENC0410 ENCON0303 ENC0320 ENC0320 ENC0307 ENC0219 ENC0408 ENC0217 ENC0417 ENC0214 ENC0406 ENC0409 ENC0319 ENC0416 ENC0315 ENC0213 ENC0417 ENC0215 ENC0309 ENC0217 ENC0405 ENC0310 ENC0314 ENC0411 ENC0205 ENC0408 ENC0420 ENC0406 ENC0317 ENC0307 ENCON0206 ENC0404 ENC0419 ENC0418 ENC0405 ENC0316 ENC0218 ENC0315 ENC0317 ENC0413 ENC0308 ENC0407 ENC0208 ENC0409 ENC0312 ENC0209 ENC0218 ENC0212 ENC0211 ENC0207 ENC0416 ENC0220 ENC0216 ENC0311 ENC0414 ENCON0204 ENC0412 ENC0420 ENC0306 ENC0412 ENC0210 ENC0216 ENC0206 ENC0312 ENC0207 ENC0311 ENC0318 ENC0209 ENC0220 ENC0305 ENC0310 ENC0318 ENCON0304 ENC0411 ENC0308 ENC0418 ENC0313 ENC0415 ENC0211 ENC0309 ENC0419 ENC0213 ENC0305 ENC0212 ENCON0203 ENC0415 ENC0407 ENC0205 ENC0304 ENC0203 ENC0404 ENC0316 ENC0303 ENC0313 ENC0306
22228-004EA 10942.56 MXN
CA89169-814 CA89169-812 89169-808 CA89169-818 CA89169-816 89169-896 89169-894 89169-812 CA89169-820 89169-898 89169-810 89169-892 89169-890 89169-816 CA89169-824 89169-814 CA89169-822 CA89169-828 CA22231-052 CA22231-050 CA89169-826 89169-818 89169-822 CA89169-832 CA89169-830 89169-820 89169-826 CA89169-836 CA89169-834 89169-824 CA22228-004 89169-828 CA89169-838 22231-052 22231-050 22228-002 89169-830 22228-004 89169-834 CA89169-842 89169-832 22228-000 CA89169-840 89169-838 CA89169-846 CA89169-844 89169-836 CA89169-848 89169-840 CA89169-850 89169-844 CA89169-854 89169-842 CA89169-852 89169-848 CA89169-858 89169-846 CA89169-856 89169-852 CA89169-860 89169-850 89169-856 CA89169-864 CA89169-862 89169-854 CA89169-902 CA89169-868 CA89169-900 CA89169-866 89169-858 CA89169-872 89169-862 CA22228-000 89169-860 CA22228-002 CA89169-870 89169-866 CA89169-876 89169-900 CA89169-874 89169-864 89169-868 CA89169-878 89169-902 CA89169-882 89169-874 CA89169-880 89169-872 89169-878 CA89169-886 CA89169-884 89169-876 89169-870 CA89169-808 CA89169-888 CA89169-894 89169-884 89169-882 CA89169-892 89169-888 CA89169-810 CA89169-898 CA89169-896 89169-886 89169-880 CA89169-890
ERGOMAT® Nitril ESD-Conductive Ergonomic Floor Mats, ERGOMAT
Alfombrillas Alfombrillas antifatiga
These mats provide a superior ergonomic matting solution in environments where the discharge of static electricity is a critical requirement.
  • Ergonomic solution in environments where discharge of static electricity is critical
  • Point-to-point resistance at 50% relative humidity is 5x104 – 1x107 ohms
  • Resistant to oils, common acids and bases in the pH 5-9 range
  • Silicone free, latex free, autoclavable at 121 °C (250 °F)
  • Nonporous, nonslip, beveled on all sides
  • Certified for use in ISO Class 5 (100) cleanrooms
  • Should not be used with vinyl-soled booties
Point to point resistance at 50% relative humidity is 5 x 104 – 1x107 ohms
Nonporous, nonslip mats offer very good chemical resistance and will not be affected by most commonly used acids and bases. Withstand chemical, steam, and high-pressure water cleaning. Constructed of virgin nitrile rubber with carbon black. Electrical resistance (point-to-point resistance at 50% RH) is 5x104–1x107 ohms. Beveled on all sides. Autoclavable at 121°C (250°F).
This mat is a perfect solution for a variety of electronics assembly and repair needs as well as for working around sensitive electronic equipment and volatile vapors and chemicals. It is resistant to oils and many common acids and bases in the pH 5-9 range.This silicone and latex free mat is also certified for use in ISO Class 5 (100) cleanrooms. Should not be used with vinyl soled booties.
Información para pedidos: ERGOMAT Nitril ESD-Conductive mats can be made in any shape and size, with every edge always beveled for safety. It has a 3 year manufacturer’s warranty when used according to manufacturer’s guidelines.