ERGOMAT® Classic Antifatigue Floor Mats, ERGOMAT®
Proveedor: ERGOMAT INC
ERGOMAT®
ST0415 ST0409 ST0218 ST0320 ST0418 ST0303 ST0411 ST0220 ST0308 ST0311 ST0319 ST0406 ST0306 ST0408 ST0214 ST0207 ST0330 ST0420 ST0311 ST0216 ST0211 ST0230 ST0206 ST0206 ST0209 ST0320 ST0260 ST0460 ST0305 ST0411 ST0318 ST0203 ST0430 ST0205 ST0209 ST0419 ST0316 ST0317 ST0416 ST0405 ST0408 ST0307 ST0306 ST0430 ST0212 ST0313 ST0215 ST0414 ST0418 ST0420 ST0406 ST0304 ST0215 ST0412 ST0412 ST0360 ST0211 ST0219 ST0207 ST0208 ST0210 ST0205 ST0220 ST0230 ST0314 ST0409 ST0315 ST0312 ST0307 ST0210 ST0417 ST0416 ST0404 ST0414 ST0404 ST0304 ST0360 ST0419 ST0305 ST0415 ST0308 ST0417 ST0219 ST0212 ST0217 ST0405 ST0313 ST0317 ST0309 ST0216 ST0204 ST0318 ST0213 ST0410 ST0208 ST0260 ST0218 ST0316 ST0407 ST0413 ST0407 ST0310 ST0203 ST0410 ST0303 ST0314 ST0319 ST0413 ST0217 ST0460 ST0213 ST0204 ST0310 ST0315 ST0330 ST0214 ST0312 ST0309
22231-010EA 4298.86 MXN
89169-092 22231-038 89169-090 22231-036 22231-034 22231-032 22231-030 89169-096 89169-094 89169-098 CA22231-108 CA22231-106 22231-108 22231-106 CA22231-116 CA22231-114 CA22231-112 CA22231-110 CA89169-058 CA89169-056 CA89169-050 CA89169-054 CA89169-052 CA22231-128 CA22231-126 CA22231-124 22231-116 22231-114 CA22231-122 CA22231-120 22231-112 22231-110 CA89169-102 CA89169-068 CA89169-100 89169-058 CA89169-066 CA89169-106 CA89169-104 89169-052 CA89169-060 89169-050 CA89169-064 89169-056 89169-054 CA89169-062 CA22231-018 22231-128 CA22231-016 22231-126 CA22231-014 22231-124 CA22231-012 CA89169-108 22231-122 CA22231-010 22231-120 CA22231-130 89169-104 89169-068 CA89169-078 89169-102 CA89169-110 89169-108 89169-106 89169-062 CA89169-072 CA89169-070 89169-060 89169-066 CA89169-076 89169-100 CA89169-074 89169-064 22231-018 CA22231-028 CA22231-027 89169-070 CA22231-026 22231-016 CA22231-024 22231-014 CA22231-022 22231-012 22231-010 CA22231-020 22231-130 CA89169-088 89169-074 CA89169-082 CA89169-080 89169-072 89169-110 89169-078 CA89169-086 89169-076 CA89169-084 CA89169-090 89169-080 CA22231-038 22231-027 CA22231-036 22231-028 22231-026 CA22231-034 CA22231-032 22231-024 22231-022 CA22231-030 22231-020 89169-084 CA89169-094 89169-082 CA89169-092 89169-088 CA89169-098 CA89169-096 89169-086
ERGOMAT® Classic Antifatigue Floor Mats, ERGOMAT®
Alfombrillas Alfombrillas antifatiga
Durable, easy to clean, and shock absorbent, these polyurethane floor mats offer outstanding ergonomic benefits
  • Durable, easy to clean, shock absorbent, static dissipative
  • For use in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanrooms
  • Ideal for dry, nonabrasive areas
  • 1.6 cm (5/8") thick with four beveled edges
Designed to reduce fatigue and combat the cumulative stress and anatomic discomforts associated with foot, leg, and back fatigue from extensive standing. Mats are static dissipative, suitable for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanrooms, and ideal for dry, nonabrasive areas. Nonslip mats are 1.6 cm (5/8") thick with four beveled edges. Available in various lengths and widths.
Información para pedidos: Manufacturer’s eight-year performance warranty accompanies these products when used in a dry, nonabrasive, noncorrosive chemical environment. Mats are available in any size or shape; contact your VWR sales representative for more information.