VWR® Mortars and Pestles, Agate
Proveedor: VWR
VWR®
C-116 C-103 C-115 C-109 C-107 C-101 C-111 C-106 C-112 C-117 C-108 C-102 C-104 C-110 C-113 C-105 C-100
89037-486CS 25252.46 MXN
89037-506 89037-498 89037-486 89037-500 89037-510 89037-488 89037-502 89037-512 89037-504 89037-514 89037-490 89037-492 89037-480 89037-494 89037-482 89037-496 89037-484
VWR® Mortars and Pestles, Agate
Morteros y manos de mortero Morteros con manos de mortero
Mortars and pestles are made of thick-walled agate, which is the most common variety of chalcedony, a semiprecious, very hard silica mineral.
  • Thick-walled agate construction
  • Highly polished surfaces
  • Octagonal shape
They feature highly polished, glossy grinding surfaces. Mortar is octagonal in shape.
Información para pedidos: Supplied in mortar and pestle sets or pestles only.