Designed for applications requiring exceptional accuracy, stability, and repeatability, VWR® Professional Hot Plate Stirrers are equipped with an exclusive safety system that helps protect both the operator and sample.
- Excellent temperature uniformity
- Digital display for temperature, speed, and time
- Cool touch, chemical-resistant housing
- Includes an RTD temperature probe kit
- Exclusive five-year warranty
- Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
- Industry Leading Safety Features
Easy-to-read touch pad controls are located on the front of the unit, with independent LED displays for temperature, speed, and time
The low-profile design occupies minimal bench space, and fits into fume hoods. The rear housing of the unit features an off-centered, built-in support rod holder with locking knob that accepts the probe kit supplied with each unit. Hot plate stirrers are available with ceramic or aluminum top plate material. White reflective ceramic tops are chemical-resistant and easy to clean. Durable aluminum tops will not crack or chip, and offer a more even heating surface.
Hot plate stirrers are equipped with a variety of innovative safety features designed to facilitate efficient operation and workplace safety. Unit housing is constructed of a heat- and chemical-resistant polymer that remains cool to the touch throughout use. The spill-resistant housing channels fluids away from internal components in the event of a spill. A maximum temperature limiting function ensures the plate temperature will not exceed preset limits, allowing for control of sensitive flash points. A hot symbol warning light is illuminated when the heat is turned on, and remains lit until the top plate is sufficiently cooled. In timed mode, an alarm will sound when time reaches zero, or when the unit reaches set point temperature. Additionally, if the probe disengages from the sample, or if the stirrer motor stops or fails, the heater will automatically shut off. Units are equipped with redundant safety controls, which will shut down unit in case of heater malfunction.
A timer displays elapsed time or a user-defined limit, and will shut off the unit when time reaches zero. The last used settings will be displayed, even after the power has been shut off. Enhanced electronics regulate the heating and stirring, and will bring samples to the desired temperature quickly and efficiently. An RTD ensures precise temperature control. A powerful continuous duty motor delivers dependable and reliable stirring at all speeds.
Certificaciones: RoHS certified. TUV listed. Calibrated Hotplate-Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the temperature and speed ranges and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Información para pedidos: Hot plate stirrers include a 234 cm (92") detachable, 3-wire cord and plug, and a 3.8 cm (11/2") PTFE coated stir bar. Units are also supplied with a probe kit that includes a 20.3 cm (8") stainless steel RTD temperature probe, 45.7 cm (18") stainless steel support rod, thermometer/temperature probe extension clamp, and hook connector. 230V, 50/60Hz models are also available; contact your VWR sales representative for more information.