VWR® Vacuum Rubber Tubing, Black
Proveedor: VWR
VWR®
09765 08591 09771 08592 09766 09767 09768 09769 09764
62993-690PK 4661.72 MXN
62993-726 62993-759 62993-690 62994-219 62994-241 62993-840 62993-817 62993-908 62993-781
VWR® Vacuum Rubber Tubing, Black
Tuberías Tuberías de vacío/presión
Tubing has especially heavy outer wall, which makes it suitable for vacuum use.
  • Seamless
  • Acid-proof
  • Good flexing qualities at low temperatures
Manufactured from good-quality natural rubber. Tubing also offers high resilience, good tensile strength, and tear resistance. Durometer hardness: Shore A, 50. Tensile strength: 2500 to 3500 psi.