VWR® Red Vacuum Rubber Tubing
Proveedor: VWR
VWR®
08596 08599 09777 09773 09775 08598 09776 09774 08597
62995-335CS 3437.97 MXN
62995-335 62995-368 62995-059 62995-426 62995-390 62995-095 62995-062 62995-153 62995-120
VWR® Red Vacuum Rubber Tubing
Tuberías Tuberías de vacío/presión
Natural rubber tubing with extra-heavy wall especially recommended for vacuum work.
  • Durometer hardness: Shore A, 50
  • Tensile strength: 2000 to 2600 psi
Has high resilience and good tensile and tear strength.