VWR® Flat Sponge Mop Head Refills
FS501V FS500V
82024-318CS 33076.8 MXN
82024-316 82024-318
VWR® Flat Sponge Mop Head Refills
Flat sponge mop refill is perfect for general use on floors, walls, and ceilings within all controlled environments.
  • Flat design
  • Suitable for general use on floors, walls, and ceilings
  • Economical refills
  • Polyester optimizes disinfectant use
Absorbs spills, cleans particulates, or applies disinfectants.
Flat sponge mop refill with polyester lamination provides tear-free application of disinfectants, cleaning, and spill absorption. Polyester allows for optimized manufacturer-recommended contact times for disinfectants. Polyester is laminated to the foam without the use of adhesives which could contaminate surfaces.
These blue refills fit both the Perfex®-style frame (14002-957) and UltraSpeed frame (89132-756). Either frame can be used with the ergonomic lightweight aluminum extendable handle (89066-060).
Información para pedidos: Mop components are sold separately.