Barrier Tapes, National Marker
Proveedor: National Marker
FBT2 BT1BY BT6RB PT40 PT30 BT2MY PT44 FBT1 PT42 BT4BY PT16 BT7RB
36434-334EA 963.04 MXN
36434-307 36434-304 36434-327 36434-305 36434-302 36434-300 36434-301 36434-334 36434-332 36434-321 36434-330 36434-308
Barrier Tapes, National Marker
Cintas
Use to mark off hazardous areas. Utilize as you would a rope.
  • Durable, lightweight polyethylene
  • Red-orange barrier tape offers 4X the visibility of other colors
  • Reusable, non-adhesive, chemical resistant, weatherproof, fade resistant
Printed barricade tapes are made of durable, lightweight polyethylene
Fluorescent red-orange barrier tape offers four times the visibility of other colors. Reusable, nonadhesive tape is resistant to most chemicals. Two-color, striped barrier tapes are made of woven polypropylene for extra durability. Chemical-resistant, weatherproof, and fade-resistant tape can be reused many times.
Printed legends warn of potential hazards and dangerous working areas.