VWR® General Purpose Cleanroom Tape, Vinyl
Proveedor: VWR
VWR®
47B-15LG 47B-1RE 47B-2DG 47B-1DB 47B-2RE 47B-3YE 47B-1TR 12YE-47B 47B-1YE 47B-2YE 47B-2LG 47B-1LB 47B-15WH 47B-34WH 47B-2BR 47B-1WH 47B-34YE 47B-12OR 47B-2WH 47B-1BL 47B-3WH 47B-12RE 47B-1DG 47B-2LB 47B-2DB 47B-1LG 47B-2OR 47B-34RE 47B-2TR 47B-12WH 47B-1OR 47B-15DB 47B-12DG 47B-2PU 47B-34DB
11211-916CS 9476.91 MXN
22221-050 22221-091 22221-092 11211-938 11211-916 11211-936 11211-914 11211-944 11211-942 11211-940 11211-962 22221-075 22221-073 75779-612 22221-077 89133-256 22221-083 22221-081 68400-834 11211-948 11211-946 11211-978 11211-956 22221-028 11211-952 11211-950 22221-086 22221-064 22221-040 22221-084 22221-062 22221-047 22221-044 22221-088 22221-067
VWR® General Purpose Cleanroom Tape, Vinyl
Cintas
VWR® General-Purpose Cleanroom Tape is ideal for use in cleanroom maintenance and support areas.
  • Multi-purpose vinyl tape
  • Color-coded for a variety of identification applications
  • 33m (36yd.) length wound on a plastic core
This multi-purpose vinyl tape is ideal for use in cleanroom maintenance and support areas. It is excellent for color coding, sealing, aisle marking, and batching. The matte finish can be written on with a cleanroom pen. Wound on a plastic core.
Información para pedidos: For additional colors and sizes, contact your Avantor representative.