VWR® Aluminum Fold-Up Hand Truck
3082
62344-934EA 4598.46 MXN
62344-934
VWR® Aluminum Fold-Up Hand Truck
Move cartons, chemicals, gas cylinders, and more with this sturdy hand truck.
  • Lightweight and collapsible for easy storage
Aluminum alloy construction
Handle extends from 71.1–106.7 cm (28–42") high. Flip-down platform size: 35.6Wx24.1Hcm (14x91/2").
Folds completely to a compact briefcase size for easy storage. Solid rubber wheels make it easy to roll. Airless tires never go flat.
Información para pedidos: Supplied with two convenient stretch cords.