Reciprocating Shaker, HS 501 digital, IKA® WORKS
2527000 2527001
33994-216EA 210021.42 MXN
33994-218 33994-216
Reciprocating Shaker, HS 501 digital, IKA® WORKS
Features a large shaking base and a compensation system to ensure quiet and stable operation
Utilizes a stepless adjustable reciprocating movement. Features a 31.8mm (11/4") stroke. Designed for loads up to 15kg (33 lbs.), including attachment. Continuous operation or timed shaking from 0 to 56 minutes. Speed digitally displayed.
Información para pedidos: Platforms sold separately. Aluminum fixing clip support 33994-266 measures 47.6Wx46Lx9.5H cm (183/4x181/8x33/4") and holds up to 50 flasks; for fixing clips see 33994-246. Dish attachment 33994-268 measures 45Wx45Lx4.4H cm (173/4x173/4x13/4") and accommodates dishes or flat bottom vessels.