Barnstead/Lab-LineThermo-Flask® Dewar Flasks, Stainless Steel, Thermo Scientific
Proveedor: Thermo Scientific
Lab-LineThermo-Flask™
2124 2123 2122 2129
63415-033EA 10987.6 MXN
63415-066 63415-033 63415-000 97049-650
Barnstead/Lab-LineThermo-Flask® Dewar Flasks, Stainless Steel, Thermo Scientific
Dewars y recipientes criogénicos
Designed for safe, effective handling and storage of all liquefied gases and other materials
Strong stainless steel containers, with sturdy handles, are rust-free, anti-corrosive and hygienic. Liners are ultra-low evacuated, high quality borosilicate glass. Manufacturer's 24-month warranty.
63415-000 , 63415-033 , 63415-066 comes with carrying handle and vented secure lid. Stainless steel lids, with self-contained insulation, fasten to container bodies with toggle clamps for safe, water-tight transporting.
97049-650 comes without lid and handle.